missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(4-methoxyphenyl)pyrazole 98.0+%, TCI America™

Catalog No. A29535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29535G 5 g
A29531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29535G Supplier TCI America Supplier No. A29535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19541-95-8
Molecular Formula C10H11N3O
Molecular Weight (g/mol) 189.218
MDL Number MFCD03093928
InChI Key UPAGEJODHNVJNM-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-1h-pyrazol-5-amine, 5-4-methoxyphenyl-1h-pyrazol-3-amine, 3-amino-5-4-methoxyphenyl pyrazole, 5-amino-3-4-methoxyphenyl pyrazole, 3-amino-5-4-methoxyphenyl-1h-pyrazole, 5-4-methoxy-phenyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methoxyphenyl-1h-pyrazole, 1h-pyrazol-3-amine, 5-4-methoxyphenyl, 5-4-methoxyphenyl-2h-pyrazol-3-amine, 5-4-methoxyphenyl-2h-pyrazol-3-ylamine
PubChem CID 2756469
IUPAC Name 5-(4-methoxyphenyl)-1H-pyrazol-3-amine
SMILES COC1=CC=C(C=C1)C2=CC(=NN2)N

Specifications

Melting Point 143°C
Color White
Quantity 5 g
Formula Weight 189.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-(4-methoxyphenyl)pyrazole