missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methoxybenzamide, 98%

Catalog No. AAA1394214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1394214 25 g
AAA1394206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1394214 Supplier Thermo Scientific Chemicals Supplier No. A1394214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17481-27-5 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17481-27-5
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017132
InChI Key INCJNDAQNPWMPZ-UHFFFAOYSA-N
Synonym benzamide, 3-amino-4-methoxy, 3-amino-p-anisamide, 3-amino-4-methoxy-benzamide, 3-amino-4-methoxy benzamide, acmc-209e8v, 5-amino-4-methoxybenzamide, cambridge id 5306680, ksc495o3j, incjndaqnpwmpz-uhfffaoysa
PubChem CID 87135
IUPAC Name 3-amino-4-methoxybenzamide
SMILES COC1=C(C=C(C=C1)C(=O)N)N

Specifications

Melting Point 156°C to 160°C
Quantity 25 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 3-Amino-4-methoxybenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-497-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only