missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-(isobutylamino)quinoline Hydrochloride 98.0+%, TCI America™

Catalog No. A27215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27215G 5 g
A27211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27215G Supplier TCI America Supplier No. A27215G

Chemical Identifiers

CAS 935521-01-0
Molecular Formula C13H18ClN3
Molecular Weight (g/mol) 251.758
MDL Number MFCD08063180
InChI Key ZRRAINCPRZTHMN-UHFFFAOYSA-N
PubChem CID 24745205
IUPAC Name 4-N-(2-methylpropyl)quinoline-3,4-diamine;hydrochloride
SMILES CC(C)CNC1=C(C=NC2=CC=CC=C21)N.Cl

Specifications

Melting Point 264°C
Quantity 5 g
Formula Weight 251.76
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-(isobutylamino)quinoline Hydrochloride