missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-hydroxyphenyl Methyl Sulfone 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 158°C |
| Quantity | 5 g |
| Formula Weight | 187.21 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Amino-4-hydroxyphenyl Methyl Sulfone |
Safety and Handling
| EINECSNumber | (3)-2140 |
| TSCA | Yes |