missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-cyclohexen-1-one, 98% (dry wt.), may cont. up to 5% water

Catalog No. AAA1147414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1147414 25 g
AAA1147406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1147414 Supplier Thermo Scientific Chemicals Supplier No. A1147414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5220-49-5 | C6H9NO | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5220-49-5
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00013783
InChI Key ZZMRPOAHZITKBV-UHFFFAOYSA-N
Synonym 3-amino-2-cyclohexen-1-one, 3-aminocyclohex-2-enone, 2-cyclohexen-1-one, 3-amino, unii-v596547zke, 3-amino-2-cyclohexen-1-ketone, 3-amino-2-cyclohexenone, 3-aminocyclohexenone, 3-aminocyclohex-enone, pubchem14449, acmc-209kyd
PubChem CID 78892
IUPAC Name 3-aminocyclohex-2-en-1-one
SMILES NC1=CC(=O)CCC1

Specifications

Melting Point 128°C to 133°C (dried)
Quantity 25 g
Beilstein 471354
Formula Weight 111.15
Percent Purity 98%
Concentration or Composition (by Analyte or Components) May contain up to 5% water
Assay (dry wt.)
Chemical Name or Material 3-Amino-2-cyclohexen-1-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-014-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only