missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2-cyclohexen-1-one, 95%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | 3-Amino-2-cyclohexen-1-one |
| Fourchette de pourcentages d’essai | 94% min. (GC) |
| Emballage | Glass bottle |
| Quantité | 5g |
| Poids de formule | 111.14 |
| Pourcentage de pureté | 95% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
| EINECSNumber | 226-014-9 |
RUO – Research Use Only