missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2-bromopyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 360600010
| Quantity | 1g |
|---|---|
| Packaging | Glass bottle |
Description
Chemical Identifiers
| 39856-58-1 | |
| 173.01 | |
| HKDVVTLISGIPFE-UHFFFAOYSA-N | |
| 642816 | |
| C1=CC(=C(N=C1)Br)N |
| C5H5BrN2 | |
| MFCD00234064 | |
| 3-amino-2-bromopyridine, 2-bromo-3-aminopyridine, 2-bromo-pyridineamine, 3-pyridinamine, 2-bromo, 2-bromo-3-pyridinamine, 2-bromo-3-pyridylamine, 2-bromopyridine-3-amine, 3-amino-2-bromo-pyridine, 2-bromo-3-pyridineamine, 2-bromo-pyridin-3-ylamine | |
| 2-bromopyridin-3-amine |
Specifications
| 3-Amino-2-bromopyridine | |
| 96.0 | |
| Authentic | |
| Glass bottle | |
| 1g | |
| 3-amino-2-bromopyridine, 2-bromo-3-aminopyridine, 2-bromo-pyridineamine, 3-pyridinamine, 2-bromo, 2-bromo-3-pyridinamine, 2-bromo-3-pyridylamine, 2-bromopyridine-3-amine, 3-amino-2-bromo-pyridine, 2-bromo-3-pyridineamine, 2-bromo-pyridin-3-ylamine | |
| HKDVVTLISGIPFE-UHFFFAOYSA-N | |
| 2-bromopyridin-3-amine | |
| 642816 | |
| 97% |
| 39856-58-1 | |
| 100.0 | |
| 96% min. (GC) | |
| C5H5BrN2 | |
| MFCD00234064 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in methanol | |
| C1=CC(=C(N=C1)Br)N | |
| 173.01 | |
| 173.01 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irritation persists: Get medi
GHS Signal Word: Danger
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.