missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-tert-butoxycarbonylpiperidine 97.0+%, TCI America™

Catalog No. A23145G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23145G 5 g
A23141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23145G Supplier TCI America Supplier No. A23145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 184637-48-7
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD01861219
InChI Key AKQXKEBCONUWCL-UHFFFAOYSA-N
Synonym 1-boc-3-aminopiperidine, n-boc-3-aminopiperidine, 3-amino-1-n-boc-piperidine, +/--3-amino-1-n-boc-piperidine, 3-amino-1-boc-piperidine, 1-n-boc-3-aminopiperidine, 3-amino-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3-amino-1-tert-butoxycarbonylpiperidine, 3-aminopiperidine-1-carboxylic acid tert-butyl ester
PubChem CID 545809
IUPAC Name tert-butyl 3-aminopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)N

Specifications

Color Yellow
Quantity 5 g
Formula Weight 200.28
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Amino-1-tert-butoxycarbonylpiperidine