missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-adamantanol 98.0+%, TCI America™

Catalog No. A20795G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20795G 5 g
A20791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20795G Supplier TCI America Supplier No. A20795G

Chemical Identifiers

CAS 702-82-9
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD01821204
InChI Key DWPIPTNBOVJYAD-UHFFFAOYSA-N
Synonym 3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
PubChem CID 658645
IUPAC Name 3-aminoadamantan-1-ol
SMILES C1C2CC3(CC1CC(C2)(C3)O)N

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 5 g
Formula Weight 167.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1-adamantanol