missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-adamantanol, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 435380010

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC435380010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

702-82-9
167.25
DWPIPTNBOVJYAD-UHFFFAOYSA-N
658645
C1C2CC3(CC1CC(C2)(C3)O)N
C10H17NO
MFCD01821204
3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
3-aminoadamantan-1-ol

Specifications

3-Amino-1-adamantanol
95.0
64.77 to 71.55% C, 7.55 to 8.35% N
Authentic
Glass bottle
1g
3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
C1C2CC3(CC1CC(C2)(C3)O)N
167.25
167.25
702-82-9
100.0
110°C
96%
C10H17NO
MFCD01821204
DWPIPTNBOVJYAD-UHFFFAOYSA-N
3-aminoadamantan-1-ol
658645
96%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions