missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzonitrile, 97+%

Catalog No. AAA1084114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1084114 25 g
AAA1084106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1084114 Supplier Thermo Scientific Chemicals Supplier No. A1084114
Only null left

CAS: 6136-68-1 | C9H7NO | 145.16 g/mol

Preparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Preparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 6136-68-1
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001806
InChI Key SBCFGFDAZCTSRH-UHFFFAOYSA-N
Synonym m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649
PubChem CID 80222
IUPAC Name 3-acetylbenzonitrile
SMILES CC(=O)C1=CC=CC(=C1)C#N

Specifications

Melting Point 97°C to 101°C
Boiling Point 120°C (5 mmHg)
Quantity 25 g
UN Number UN3439
Beilstein 2079629
Solubility Information Insoluble in water.
Formula Weight 145.16
Percent Purity ≥97%
Chemical Name or Material 3-Acetylbenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 228-110-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only