missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2,4-dimethylpyrrole, 98%, Thermo Scientific Chemicals

Catalog No. AC102510500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102510500 50 g Glass bottle
AC102510100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102510500 Supplier Thermo Scientific Chemicals Supplier No. 102510500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2386-25-6
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
InChI Key VGZCKCJMYREIKA-UHFFFAOYSA-N
Synonym 3-acetyl-2,4-dimethylpyrrole, 2,4-dimethyl-3-acetylpyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl ethanone, 2,4-dimethylpyrrol-3-yl methyl ketone, ethanone, 1-2,4-dimethyl-1h-pyrrol-3-yl, ketone, 2,4-dimethylpyrrol-3-yl methyl, 1-2,4-dimethyl-1h-pyrrol-3-yl ethan-1-one, 2,4-dimethyl-3-acetyl-pyrrole, 3-acetyl-2,5-dimethyl-pyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl-ethanone
PubChem CID 15163
IUPAC Name 1-(2,4-dimethyl-1H-pyrrol-3-yl)ethanone
SMILES CC1=CNC(=C1C(=O)C)C

Specifications

Melting Point 136.0°C to 139.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Formula Weight 137.18
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 3-Acetyl-2, 4-dimethylpyrrole
Hazard Category GHS Signal Word: Warning
EINECSNumber 219-197-1

RUO – Research Use Only