missing translation for 'onlineSavingsMsg'
Learn More

3'-Acetamidoacetophenone 98.0+%, TCI America™

Catalog No. A1408500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1408500G 500 g
A140825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1408500G Supplier TCI America Supplier No. A1408500G

Chemical Identifiers

CAS 7463-31-2
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD00032278
InChI Key AFZTYHRVDOKRKV-UHFFFAOYSA-N
Synonym 3'-acetamidoacetophenone, n-3-acetylphenyl acetamide, 3'-n-acetylamino acetophenone, 3'-acetylacetanilide, 3-acetamidoacetophenone, 3-acetamido acetophenone, m-acetylaminoacetophenone, 3-acetylacetanilide, acetamide, n-3-acetylphenyl
PubChem CID 346202
IUPAC Name N-(3-acetylphenyl)acetamide
SMILES CC(=O)C1=CC(=CC=C1)NC(=O)C

Specifications

Melting Point 127°C
Quantity 500 g
Formula Weight 177.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Acetamidoacetophenone