missing translation for 'onlineSavingsMsg'
Learn More

3,9-Diphenyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 98.0+%, TCI America™

Catalog No. p-7128633
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5056200MG 200 mg
D50561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5056200MG Supplier TCI America Supplier No. D5056200MG

Chemical Identifiers

CAS 1359833-28-5
Molecular Formula C30H28BNO2
Molecular Weight (g/mol) 445.37
MDL Number MFCD29089360
InChI Key QVMVGEABPNCFOY-UHFFFAOYSA-N
Synonym 6,9-Diphenylcarbazole-3-boronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-(6,9-diphenylcarbazol-3-yl)-1,3,2-dioxaborolane
PubChem CID 71143518
IUPAC Name 3,9-diphenyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
SMILES CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)N(C1=C2C=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164°C
Color White
Quantity 200 mg
Formula Weight 445.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,9-Diphenyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole