missing translation for 'onlineSavingsMsg'
Learn More

3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane 95.0+%, TCI America™

Catalog No. p-7124153
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56645G 5 g
B566425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56645G Supplier TCI America Supplier No. B56645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26741-53-7
Molecular Formula C33H50O6P2
Molecular Weight (g/mol) 604.71
MDL Number MFCD00071521
InChI Key AIBRSVLEQRWAEG-FJGSWBKINA-N
Synonym Pentaerythritol Bis(2,4-di-tert-butylphenylphosphite)
PubChem CID 93101
IUPAC Name 3,9-bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
SMILES CC(C)(C)C1=CC=C(O[P@]2OC[C@]3(CO[P@](OC4=CC=C(C=C4C(C)(C)C)C(C)(C)C)OC3)CO2)C(=C1)C(C)(C)C

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
Formula Weight 604.70
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
EINECSNumber (5)-5768
TSCA Yes