missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dimethyl-1-octanol 98.0+%, TCI America™

Catalog No. D144225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D144225ML 25 mL
D1442500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D144225ML Supplier TCI America Supplier No. D144225ML

Chemical Identifiers

CAS 106-21-8
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00002936
InChI Key PRNCMAKCNVRZFX-UHFFFAOYSA-N
Synonym 3,7-dimethyl-1-octanol, tetrahydrogeraniol, dimethyloctanol, dihydrocitronellol, pelargol, perhydrogeraniol, geraniol tetrahydride, citronellol, dihydro, 1-octanol, 3,7-dimethyl, geraniol, perhydro
PubChem CID 7792
IUPAC Name 3,7-dimethyloctan-1-ol
SMILES CC(C)CCCC(C)CCO

Specifications

Color Colorless
Boiling Point 118°C
Quantity 25 mL
Formula Weight 158.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,7-Dimethyl-1-octanol
EINECSNumber (2)-0217
RTECSNumber RH0900000
TSCA Yes