missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Thioxanthenediamine-10,10-dioxide, 97%, Thermo Scientific™
$954.59
Chemical Identifiers
| CAS | 10215-25-5 |
|---|---|
| Molecular Formula | C13H12N2O2S |
| Molecular Weight (g/mol) | 260.32 |
| MDL Number | MFCD00041841 |
| InChI Key | UPVRZVIJGVFROW-UHFFFAOYSA-N |
| Synonym | 9h-thioxanthene-3,6-diamine 10,10-dioxide, 9h-thioxanthene-3,6-diamine, 10,10-dioxide, 3,6-diamino-9h-thioxanthene 10,10-dioxide, 3,6-thioxanthenediamine-10,10-dioxide, 3,7-thiaxanthenediamine-5,5-dioxide, 9h-thioxanthene-3,7-diamine, 5,5-dioxide, 9h-thioxanthene-3,6-diamine,10,10-dioxide, 3,7-diamino-10h-dibenzo b,e thiin-5,5-dione, 3,6-diamino-9h-10??-thioxanthene-10,10-dione, 3,6-diamino-9h-10 |
| PubChem CID | 82456 |
| IUPAC Name | 10,10-dioxo-9H-thioxanthene-3,6-diamine |
| SMILES | C1C2=C(C=C(C=C2)N)S(=O)(=O)C3=C1C=CC(=C3)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC420910010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC420910100
|
N/A | 10g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 10215-25-5 | |
| 260.32 | |
| UPVRZVIJGVFROW-UHFFFAOYSA-N | |
| 82456 | |
| C1C2=C(C=C(C=C2)N)S(=O)(=O)C3=C1C=CC(=C3)N |
| C13H12N2O2S | |
| MFCD00041841 | |
| 9h-thioxanthene-3,6-diamine 10,10-dioxide, 9h-thioxanthene-3,6-diamine, 10,10-dioxide, 3,6-diamino-9h-thioxanthene 10,10-dioxide, 3,6-thioxanthenediamine-10,10-dioxide, 3,7-thiaxanthenediamine-5,5-dioxide, 9h-thioxanthene-3,7-diamine, 5,5-dioxide, 9h-thioxanthene-3,6-diamine,10,10-dioxide, 3,7-diamino-10h-dibenzo b,e thiin-5,5-dione, 3,6-diamino-9h-10??-thioxanthene-10,10-dione, 3,6-diamino-9h-10 | |
| 10,10-dioxo-9H-thioxanthene-3,6-diamine |
Specifications
| 10215-25-5 | |
| 100.0 | |
| 96% min. (HPLC) | |
| MFCD00041841 | |
| UPVRZVIJGVFROW-UHFFFAOYSA-N | |
| 10,10-dioxo-9H-thioxanthene-3,6-diamine | |
| 82456 | |
| 97% |
| 96.0 | |
| Authentic | |
| C13H12N2O2S | |
| 9h-thioxanthene-3,6-diamine 10,10-dioxide, 9h-thioxanthene-3,6-diamine, 10,10-dioxide, 3,6-diamino-9h-thioxanthene 10,10-dioxide, 3,6-thioxanthenediamine-10,10-dioxide, 3,7-thiaxanthenediamine-5,5-dioxide, 9h-thioxanthene-3,7-diamine, 5,5-dioxide, 9h-thioxanthene-3,6-diamine,10,10-dioxide, 3,7-diamino-10h-dibenzo b,e thiin-5,5-dione, 3,6-diamino-9h-10??-thioxanthene-10,10-dione, 3,6-diamino-9h-10 | |
| C1C2=C(C=C(C=C2)N)S(=O)(=O)C3=C1C=CC(=C3)N | |
| 260.32 | |
| 260.32 | |
| 3, 6-Thioxanthenediamine-10, 10-dioxide, 97% |
Safety and Handling
EINECSNumber : 233-527-1
RTECSNumber : XO0668000
TSCA : TSCA
RUO – Research Use Only