accessibility menu, dialog, popup

UserName

CAS RN 10215-25-5

IUPAC Name: 3,6-diamino-9H-10λ⁶-thioxanthene-10,10-dione
Synonyms: 3,6-diamino-9H-10λ⁶-thioxanthene-10,10-dione
Molecular Weight (g/mol): 260.31
Molecular Formula: C13H12N2O2S
InChi Key: UPVRZVIJGVFROW-UHFFFAOYSA-N
SMILES: NC1=CC=C2CC3=CC=C(N)C=C3S(=O)(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results