missing translation for 'onlineSavingsMsg'
Learn More

3,6-Thioxanthenediamine-10,10-dioxide, 97%

Catalog No. AC420910100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420910100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420910100 Supplier Thermo Scientific Chemicals Supplier No. 420910100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10215-25-5 | C13H12N2O2S | 260.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10215-25-5
Molecular Formula C13H12N2O2S
Molecular Weight (g/mol) 260.32
MDL Number MFCD00041841
InChI Key UPVRZVIJGVFROW-UHFFFAOYSA-N
PubChem CID 82456
IUPAC Name 10,10-dioxo-9H-thioxanthene-3,6-diamine
SMILES C1C2=C(C=C(C=C2)N)S(=O)(=O)C3=C1C=CC(=C3)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 213°C to 215°C
Color Beige to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Synonym 9h-thioxanthene-3,6-diamine 10,10-dioxide, 9h-thioxanthene-3,6-diamine, 10,10-dioxide, 3,6-diamino-9h-thioxanthene 10,10-dioxide, 3,6-thioxanthenediamine-10,10-dioxide, 3,7-thiaxanthenediamine-5,5-dioxide, 9h-thioxanthene-3,7-diamine, 5,5-dioxide, 9h-thioxanthene-3,6-diamine,10,10-dioxide, 3,7-diamino-10h-dibenzo b,e thiin-5,5-dione, 3,6-diamino-9h-10??-thioxanthene-10,10-dione, 3,6-diamino-9h-10
Formula Weight 260.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3, 6-Thioxanthenediamine-10, 10-dioxide, 97%
Show More Show Less
EINECSNumber 233-527-1
RTECSNumber XO0668000
TSCA TSCA

RUO – Research Use Only