missing translation for 'onlineSavingsMsg'
Learn More

3,6-Thioxanthenediamine-10,10-dioxide, 97%, Thermo Scientific™

Catalog No. AC420910010
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC420910010 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420910010 Supplier Thermo Scientific Chemicals Supplier No. 420910010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 10215-25-5
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C13H12N2O2S
Quantity 1g
MDL Number MFCD00041841
Synonym 9h-thioxanthene-3,6-diamine 10,10-dioxide, 9h-thioxanthene-3,6-diamine, 10,10-dioxide, 3,6-diamino-9h-thioxanthene 10,10-dioxide, 3,6-thioxanthenediamine-10,10-dioxide, 3,7-thiaxanthenediamine-5,5-dioxide, 9h-thioxanthene-3,7-diamine, 5,5-dioxide, 9h-thioxanthene-3,6-diamine,10,10-dioxide, 3,7-diamino-10h-dibenzo b,e thiin-5,5-dione, 3,6-diamino-9h-10??-thioxanthene-10,10-dione, 3,6-diamino-9h-10
InChI Key UPVRZVIJGVFROW-UHFFFAOYSA-N
SMILES C1C2=C(C=C(C=C2)N)S(=O)(=O)C3=C1C=CC(=C3)N
IUPAC Name 10,10-dioxo-9H-thioxanthene-3,6-diamine
Molecular Weight (g/mol) 260.32
PubChem CID 82456
Formula Weight 260.32
Percent Purity 97%
Chemical Name or Material 3, 6-Thioxanthenediamine-10, 10-dioxide, 97%
Show More Show Less
EINECSNumber 233-527-1
RTECSNumber XO0668000
TSCA TSCA

RUO – Research Use Only