missing translation for 'onlineSavingsMsg'
Learn More

{3-[(6-Methylpyrazin-2-yl)oxy]phenyl}methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 906352-98-5
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD09817525
InChI Key DWEAUGVLWACZHO-UHFFFAOYSA-N
Synonym 3-6-methylpyrazin-2-yl oxy phenyl methanol, 3-6-methylpyrazin-2-yloxy phenyl methan-1-ol, benzenemethanol,3-6-methyl-2-pyrazinyl oxy
PubChem CID 24229679
IUPAC Name [3-(6-methylpyrazin-2-yl)oxyphenyl]methanol
SMILES CC1=CN=CC(=N1)OC2=CC=CC(=C2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC62609CB
SDS View Documents
Thermo Scientific Chemicals
CC62609CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC62609DA
SDS View Documents
Thermo Scientific Chemicals
CC62609DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

906352-98-5
216.24
DWEAUGVLWACZHO-UHFFFAOYSA-N
24229679
CC1=CN=CC(=N1)OC2=CC=CC(=C2)CO
C12H12N2O2
MFCD09817525
3-6-methylpyrazin-2-yl oxy phenyl methanol, 3-6-methylpyrazin-2-yloxy phenyl methan-1-ol, benzenemethanol,3-6-methyl-2-pyrazinyl oxy
[3-(6-methylpyrazin-2-yl)oxyphenyl]methanol

Specifications

906352-98-5
100.0
Amber Glass Bottle
MFCD09817525
3-6-methylpyrazin-2-yl oxy phenyl methanol, 3-6-methylpyrazin-2-yloxy phenyl methan-1-ol, benzenemethanol,3-6-methyl-2-pyrazinyl oxy
CC1=CN=CC(=N1)OC2=CC=CC(=C2)CO
216.24
216.24
{3-[(6-Methylpyrazin-2-yl)oxy]phenyl}methanol
96.5
97%
C12H12N2O2
250mg
DWEAUGVLWACZHO-UHFFFAOYSA-N
[3-(6-methylpyrazin-2-yl)oxyphenyl]methanol
24229679
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents