missing translation for 'onlineSavingsMsg'
Learn More

3-[(6-Methylpyrazin-2-yl)oxy]aniline, 97%, Thermo Scientific™

Catalog No. CC62614CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC62614CB 250mg Amber Glass Bottle
CC62614DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC62614CB Supplier Thermo Scientific Chemicals Supplier No. CC62614CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 915707-61-8
Molecular Formula C11H11N3O
Molecular Weight (g/mol) 201.229
MDL Number MFCD09702406
InChI Key ZFDWDKWQAXTCSU-UHFFFAOYSA-N
Synonym 3-6-methylpyrazin-2-yl oxy aniline, benzenamine, 3-6-methyl-2-pyrazinyl oxy, 3-6-methylpyrazin-2-yloxy phenylamine, benzenamine,3-6-methyl-2-pyrazinyl oxy
PubChem CID 24229682
IUPAC Name 3-(6-methylpyrazin-2-yl)oxyaniline
SMILES CC1=CN=CC(=N1)OC2=CC=CC(=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 201.23
Percent Purity 97%
Chemical Name or Material 3-[(6-Methylpyrazin-2-yl)oxy]aniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.