missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dithia-1,8-octanediol, 97%
CAS: 5244-34-8 | C6H14O2S2 | 182.296 g/mol
Supplier: Thermo Scientific Chemicals A1765418
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3,6-Dithia-1,8-octanediol | |
| 63°C to 67°C | |
| C6H14O2S2 | |
| 50 g | |
| 3,6-dithia-1,8-octanediol, 1,2-bis 2-hydroxyethylthio ethane, 3,6-dithiaoctane-1,8-diol, dithiaoctanediol, ethylenedithioethanol, ethanol, 2,2'-1,2-ethanediylbis thio bis, unii-kbc80263g6, 2,2'-ethane-1,2-diylbis sulfanediyl diethanol, 2,2'-ethylenedithio diethanol, ethanol, 2,2'-ethylenedithio di | |
| C(CSCCSCCO)O | |
| 182.296 | |
| 182.31 |
| 5244-34-8 | |
| 170°C (0.8 mmHg) | |
| MFCD00002911 | |
| 1739193 | |
| PDHFSBXFZGYBIP-UHFFFAOYSA-N | |
| 2-[2-(2-hydroxyethylsulfanyl)ethylsulfanyl]ethanol | |
| 78904 | |
| 97% |
Chemical Identifiers
| 5244-34-8 | |
| 182.296 | |
| PDHFSBXFZGYBIP-UHFFFAOYSA-N | |
| 78904 | |
| C(CSCCSCCO)O |
| C6H14O2S2 | |
| MFCD00002911 | |
| 3,6-dithia-1,8-octanediol, 1,2-bis 2-hydroxyethylthio ethane, 3,6-dithiaoctane-1,8-diol, dithiaoctanediol, ethylenedithioethanol, ethanol, 2,2'-1,2-ethanediylbis thio bis, unii-kbc80263g6, 2,2'-ethane-1,2-diylbis sulfanediyl diethanol, 2,2'-ethylenedithio diethanol, ethanol, 2,2'-ethylenedithio di | |
| 2-[2-(2-hydroxyethylsulfanyl)ethylsulfanyl]ethanol |
Safety and Handling
EINECSNumber : 226-045-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only