accessibility menu, dialog, popup

UserName

HOSSOH

CAS RN 5244-34-8

IUPAC Name: 2-({2-[(2-hydroxyethyl)sulfanyl]ethyl}sulfanyl)ethan-1-ol
Synonyms: dithiaoctanediol
Molecular Weight (g/mol): 182.3
Molecular Formula: C6H14O2S2
InChi Key: PDHFSBXFZGYBIP-UHFFFAOYSA-N
SMILES: OCCSCCSCCO
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)