missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dithia-1,8-octanediol, 97%

Catalog No. p-7023770
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1765409 10 g
AAA1765418 50 g
AAA1765430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1765409 Supplier Thermo Scientific Chemicals Supplier No. A1765409

CAS: 5244-34-8 | C6H14O2S2 | 182.296 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5244-34-8
Molecular Formula C6H14O2S2
Molecular Weight (g/mol) 182.296
MDL Number MFCD00002911
InChI Key PDHFSBXFZGYBIP-UHFFFAOYSA-N
Synonym 3,6-dithia-1,8-octanediol, 1,2-bis 2-hydroxyethylthio ethane, 3,6-dithiaoctane-1,8-diol, dithiaoctanediol, ethylenedithioethanol, ethanol, 2,2'-1,2-ethanediylbis thio bis, unii-kbc80263g6, 2,2'-ethane-1,2-diylbis sulfanediyl diethanol, 2,2'-ethylenedithio diethanol, ethanol, 2,2'-ethylenedithio di
PubChem CID 78904
IUPAC Name 2-[2-(2-hydroxyethylsulfanyl)ethylsulfanyl]ethanol
SMILES C(CSCCSCCO)O

Specifications

Melting Point 63°C to 67°C
Boiling Point 170°C (0.8 mmHg)
Quantity 10 g
Beilstein 1739193
Formula Weight 182.31
Percent Purity 97%
Chemical Name or Material 3,6-Dithia-1,8-octanediol
EINECSNumber 226-045-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only