missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydroxyflavone 98.0+%, TCI America™

Catalog No. D51911G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D51911G Supplier TCI America Supplier No. D51911G
Only null left

Chemical Identifiers

CAS 108238-41-1
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00143082
InChI Key XHLOLFKZCUCROE-UHFFFAOYSA-N
Synonym 3,6-dihydroxyflavone, 3,6-dihydroxy-2-phenyl-4h-chromen-4-one, 6-hydroxyflavonol, 3,6-dihydroxy-flavone, 4h-1-benzopyran-4-one, 3,6-dihydroxy-2-phenyl, acmc-1c7fz, 3,6-dihydroxy-2-phenylchromone
PubChem CID 688659
IUPAC Name 3,6-dihydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=C(C(=O)C3=C(O2)C=CC(=C3)O)O

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 1 g
Formula Weight 254.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dihydroxyflavone