missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloropyridazine, 98%

Catalog No. AAA1479514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1479514 25 g
AAA1479522 100 g
AAA1479536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1479514 Supplier Thermo Scientific Chemicals Supplier No. A1479514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 141-30-0 | C4H2Cl2N2 | 148.974 g/mol

3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 141-30-0
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00006466
InChI Key GUSWJGOYDXFJSI-UHFFFAOYSA-N
Synonym pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine
PubChem CID 67331
IUPAC Name 3,6-dichloropyridazine
SMILES C1=CC(=NN=C1Cl)Cl

Specifications

Melting Point 66°C to 70°C
Quantity 25 g
UN Number UN2811
Beilstein 111151
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 148.98
Percent Purity 98%
Chemical Name or Material 3,6-Dichloropyridazine
Hazard Category H301-H315-H319-H332-H335
Hazard Statement GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 205-478-6
RTECSNumber UR5886250
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only