missing translation for 'onlineSavingsMsg'
Learn More

3',6'-Bis(diethylamino)-2-(4-nitrophenyl)spiro[isoindole-1,9'-xanthene]-3-one 98.0+%, TCI America™

Catalog No. B262825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B262825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B262825G Supplier TCI America Supplier No. B262825G
Only null left

Chemical Identifiers

CAS 29199-09-5
Molecular Formula C34H34N4O4
Molecular Weight (g/mol) 562.67
InChI Key XZXFZILEZWXEND-UHFFFAOYSA-N
PubChem CID 120150
IUPAC Name 3',6'-bis(diethylamino)-2-(4-nitrophenyl)spiro[isoindole-3,9'-xanthene]-1-one
SMILES CCN(CC)C1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)N(CC)CC)C5=CC=CC=C5C(=O)N3C6=CC=C(C=C6)[N+](=O)[O-]

Specifications

Melting Point 200°C
Quantity 25 g
Formula Weight 562.67
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3′,6′-Bis(diethylamino)-2-(4-nitrophenyl)spiro[isoindole-1,9′-xanthene]-3-one
EINECSNumber (5)-3639
TSCA No