missing translation for 'onlineSavingsMsg'
Learn More

3,5-Heptanedione 97.0+%, TCI America™

Catalog No. H13955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13955G 5 g
H139525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13955G Supplier TCI America Supplier No. H13955G

Chemical Identifiers

CAS 7424-54-6
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00015186
InChI Key DGCTVLNZTFDPDJ-UHFFFAOYSA-N
Synonym 3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc
PubChem CID 81923
IUPAC Name heptane-3,5-dione
SMILES CCC(=O)CC(=O)CC

Specifications

Color Yellow
Boiling Point 177°C
Quantity 5 g
UN Number 1224
Formula Weight 128.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Heptanedione
EINECSNumber (2)-3141
TSCA Yes