missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diphenylpyrazole 98.0+%, TCI America™

Catalog No. D41975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41975G 5 g
D419725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41975G Supplier TCI America Supplier No. D41975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1145-01-3
Molecular Formula C15H12N2
Molecular Weight (g/mol) 220.28
MDL Number MFCD00039675
InChI Key JXHKUYQCEJILEI-UHFFFAOYSA-N
Synonym 3,5-diphenylpyrazole, 1h-pyrazole, 3,5-diphenyl, pyrazole, 3,5-diphenyl, pyrazole,5-diphenyl, acmc-1bufo, 1h-pyrazole,5-diphenyl, cbmicro_048085, 3,5-diphenyl pyrazole, 3,5-diphenyl-1h-pyrazole 5g
PubChem CID 70840
IUPAC Name 3,5-diphenyl-1H-pyrazole
SMILES N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 202°C
Color Yellow
Quantity 5 g
Formula Weight 220.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diphenylpyrazole