missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoyl chloride, 98+%

Catalog No. AAA1672922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1672922 100 g
AAA1672914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1672922 Supplier Thermo Scientific Chemicals Supplier No. A1672922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-33-2 | C7H3ClN2O5 | 230.56 g/mol

Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water.

Chemical Identifiers

CAS 99-33-2
Molecular Formula C7H3ClN2O5
Molecular Weight (g/mol) 230.56
MDL Number MFCD00007248
InChI Key NNOHXABAQAGKRZ-UHFFFAOYSA-N
Synonym benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc;
PubChem CID 7432
IUPAC Name 3,5-dinitrobenzoyl chloride
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl

Specifications

Melting Point 67°C to 70°C
Boiling Point 196°C (12 mmHg)
Quantity 100 g
UN Number UN3261
Beilstein 990249
Sensitivity Moisture sensitive
Merck Index 14,3277
Solubility Information Slightly soluble in water.
Formula Weight 230.57
Percent Purity ≥98%
Chemical Name or Material 3,5-Dinitrobenzoyl chloride
Show More Show Less
Hazard Category H314-H341
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P260-P264b-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 202-750-6
RTECSNumber DM6637000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only