missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitroaniline 98.0+%, TCI America™

Catalog No. D24585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24585G 5 g
D245825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24585G Supplier TCI America Supplier No. D24585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 618-87-1
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.123
MDL Number MFCD00007263
InChI Key MPBZUKLDHPOCLS-UHFFFAOYSA-N
PubChem CID 12068
IUPAC Name 3,5-dinitroaniline
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])N

Specifications

Melting Point 162°C
Quantity 5 g
UN Number 1596
Synonym benzenamine, 3,5-dinitro, aniline, 3,5-dinitro, unii-0ahr6k1n73, ccris 3109, 3,5-dinitrophenylamine, 3-5-dinitro-phenylamine, 3,5-dinitroanilin, aniline,5-dinitro, acmc-209mx8, benzenamine,3,5-dinitro
Formula Weight 183.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dinitroaniline
RTECSNumber BX9200100
TSCA No