missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenol 98.0+%, TCI America™

Catalog No. D182925G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D182925G 25 g
D1829100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D182925G Supplier TCI America Supplier No. D182925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-68-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002307
InChI Key TUAMRELNJMMDMT-UHFFFAOYSA-N
Synonym 3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa
PubChem CID 7948
ChEBI CHEBI:38572
IUPAC Name 3,5-dimethylphenol
SMILES CC1=CC(O)=CC(C)=C1

Specifications

Melting Point 64°C
Boiling Point 95°C
Quantity 25 g
UN Number 2261
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethylphenol
EINECSNumber (3)-0521&(4)-0057
RTECSNumber ZE6475000
TSCA Yes