missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenol, 99+%

Catalog No. AC162795000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162795000 500 g Glass bottle
AC162790050 5 g Glass bottle
AC162791000 100 g Glass bottle
AC162790025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC162795000 Supplier Thermo Scientific Chemicals Supplier No. 162795000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-68-9 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-68-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002307
InChI Key TUAMRELNJMMDMT-UHFFFAOYSA-N
Synonym 3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa
PubChem CID 7948
ChEBI CHEBI:38572
IUPAC Name 3,5-dimethylphenol
SMILES CC1=CC(O)=CC(C)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 63°C to 66°C
Color Orange to White
Density 1.1150g/mL
Boiling Point 222°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C6H3OH
Quantity 500 g
Beilstein 06, 492
Specific Gravity 1.115
Merck Index 15, 10279
Solubility Information Solubility in water: 5.3g/L (25°C). Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene
Formula Weight 122.17
Percent Purity 99+%
Physical Form Crystalline Solid
Chemical Name or Material 3, 5-Dimethylphenol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-606-5
RTECSNumber ZE6475000
TSCA TSCA

RUO – Research Use Only