missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC00601EB
Change view
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00601EB 25g Amber Glass Bottle
CC00601DA 1g Amber Glass Bottle
CC00601DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC00601EB Supplier Thermo Scientific Chemicals Supplier No. CC00601EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2510-36-3
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
InChI Key IJEUISLJVBUNRE-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole-4-carboxylic acid, 3,5-dimethylisoxasole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3,5-dimethyl, dimethyl-1,2-oxazole-4-carboxylic acid, 3,5-dimethyl-4-isoxazolecarboxylic acid, 3,5-dimethyl-isoxazole-4-carboxylic acid, 3,5-dimethylisoxazole-4-carboxylicacid, pubchem8774, akos pao-1555
PubChem CID 75636
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(=NO1)C)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25g
Formula Weight 141.13
Chemical Name or Material 3, 5-Dimethylisoxazole-4-carboxylic acid