missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonitrile, ≥97%, Thermo Scientific™

Numéro de catalogue. CC48016DA
Modifier l'affichage
Click to view available options
Quantité:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC48016DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC48016DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC48016DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 23198-55-2
Formule moléculaire C12H11N3
Poids moléculaire (g/mol) 197.241
Numéro MDL MFCD08435888
Clé InChI WOFAIVGOIVCZJJ-UHFFFAOYSA-N
Synonyme 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile,3,5-dimethyl-1-phenyl
PubChem CID 12208701
Nom de l’IUPAC 3,5-dimethyl-1-phenylpyrazole-4-carbonitrile
SOURIRES CC1=C(C(=NN1C2=CC=CC=C2)C)C#N

Spécifications

Nom ou substance chimique 3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonitrile
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 1g
Poids de formule 197.24
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.