missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxytoluene, 98%

Catalog No. AAL0080914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0080914 25 g
AAL0080903 1 g
AAL0080906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0080914 Supplier Thermo Scientific Chemicals Supplier No. L0080914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4179-19-5 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4179-19-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00015435
InChI Key RIZBLVRXRWHLFA-UHFFFAOYSA-N
Synonym 3,5-dimethoxytoluene, orcinol dimethyl ether, benzene, 1,3-dimethoxy-5-methyl, 5-methylresorcinol dimethyl ether, 3,5-dimethoxy toluene, toluene, 3,5-dimethoxy, 1,3-dimethoxy-5-methyl-benzene, pubchem17775, acmc-1ak5i
PubChem CID 77844
IUPAC Name 1,3-dimethoxy-5-methylbenzene
SMILES COC1=CC(OC)=CC(C)=C1

Specifications

Density 1.039
Boiling Point 244°C
Flash Point 101°C (213°F)
Odor Aromatic
Refractive Index 1.5225
Quantity 25 g
Beilstein 2043558
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxytoluene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-048-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only