missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenylacetonitrile, 98+%

Catalog No. AAL1144306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1144306 5 g
AAL1144303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1144306 Supplier Thermo Scientific Chemicals Supplier No. L1144306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13388-75-5 | C10H11NO2 | 177.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13388-75-5
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00016395
InChI Key UUNRWZQWCNTSCV-UHFFFAOYSA-N
Synonym 3,5-dimethoxyphenylacetonitrile, 2-3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxy-benzeneacetonitrile, benzeneacetonitrile, 3,5-dimethoxy, 2-3,5-dimethoxyphenyl ethanenitrile, pubchem16207, 3,5-dimethoxybenzylcyanide, acmc-20a41u
PubChem CID 139445
IUPAC Name 2-(3,5-dimethoxyphenyl)acetonitrile
SMILES COC1=CC(OC)=CC(CC#N)=C1

Specifications

Melting Point 53°C to 56°C
Odor Characteristic
Quantity 5 g
Beilstein 2096481
Formula Weight 177.2
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethoxyphenylacetonitrile
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only