Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,5-Diméthoxyphénol, 97%
Description
Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de produits d’Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original d’Acros Organics ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.
Specifications
Specifications
| Nom ou substance chimique | 3, 5-Dimethoxyphenol |
| CAS % minimum | 96.0 |
| CAS % maximal | 100.0 |
| Point de fusion | 40 °C à 45 °C |
| Couleur | Brun-rosé à incolore |
| Point d’ébullition | 172 °C à 175 °C (17,0 mmHg) |
| Point d’éclair | 78 °C |
| Spectre infrarouge | Authentique |
| Fourchette de pourcentages d’essai | 96% min. (GC) |
| Emballage | Bouteille en verre |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 207-917-7 |
Utilisation uniquement de la recherche RUO –