missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzylamine, 98%

Catalog No. AAH6050306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6050306 5 g
AAH6050314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6050306 Supplier Thermo Scientific Chemicals Supplier No. H6050306

CAS: 34967-24-3 | C9H13NO2 | 167.208 g/mol

3,5-Dimethoxybenzylamine was used in the preparation of trisammonium tris(hexafluoro phosphate) salt.  It was also used in the synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxybenzylamine was used in the preparation of trisammonium tris(hexafluoro phosphate) salt.  It was also used in the synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.

Chemical Identifiers

CAS 34967-24-3
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00052813
InChI Key YGZJTYCCONJJGZ-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzylamine, 3,5-dimethoxybenzyl amine, 3,5-dimethoxyphenyl methanamine, 1-3,5-dimethoxyphenyl methanamine, 3,5-dimethoxy benzyl amine, 3,5-dimethoxyphenyl methylamine, benzenemethanamine, 3,5-dimethoxy, pubchem7393, 3,5-dimethoxy benzylamine, 3,5-dimethoxy-benzylamine
PubChem CID 420973
IUPAC Name (3,5-dimethoxyphenyl)methanamine
SMILES COC1=CC(=CC(=C1)CN)OC

Specifications

Melting Point 35°C to 37°C
Density 1.106
Boiling Point 94°C to 96°C
Flash Point 113°C (235°F)
Refractive Index 1.542
Quantity 5 g
UN Number UN3259
Solubility Information Slightly soluble in water.
Formula Weight 167.21
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxybenzylamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only