missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzoic acid, 97%

Catalog No. AC114905000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC114905000 500 g
AC114901000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC114905000 Supplier Thermo Scientific Chemicals Supplier No. 114905000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-10-5 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-10-5
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002512
InChI Key UYEMGAFJOZZIFP-UHFFFAOYSA-N
Synonym alpha-resorcylic acid, 5-carboxyresorcinol, benzoic acid, 3,5-dihydroxy, unii-2wc5lmo6l1, 3,5-dihydroxy benzoic acid, 3,5-dihydroxy-benzoic acid, 3,5-dhba, 3,5-dihydroxybenzoicacid, a-resorcylic acid, .alpha.-resorcylic acid
PubChem CID 7424
ChEBI CHEBI:39912
IUPAC Name 3,5-dihydroxybenzoic acid
SMILES OC(=O)C1=CC(O)=CC(O)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 235.0°C to 240.0°C
Color White to Beige
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula (HO)2C6H3CO2H
Quantity 500 g
Beilstein 10,404
Solubility Information Solubility in water: 84g/L (20°C). Other solubilities: soluble in ethanol and ether
Formula Weight 154.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3,5-Dihydroxybenzoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-730-7
RTECSNumber VH3708000
TSCA TSCA

RUO – Research Use Only