missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorosalicylic acid, 98+%, Thermo Scientific Chemicals

Catalog No. AAH3294003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3294003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3294003 Supplier Thermo Scientific Chemicals Supplier No. H3294003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 84376-20-5 | Molecular Formula: C7H4F2O3 | Molecular Weight: 174.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84376-20-5
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.103
MDL Number MFCD06203609
InChI Key GZPCNALAXFNOBT-UHFFFAOYSA-N
Synonym 3,5-difluorosalicylic acid, 3,5-difluoro-salicylic acid, benzoic acid, 3,5-difluoro-2-hydroxy
PubChem CID 10888405
IUPAC Name 3,5-difluoro-2-hydroxybenzoic acid
SMILES C1=C(C=C(C(=C1F)O)C(=O)O)F

Specifications

Melting Point 187°C to 191°C
Quantity 1 g
Formula Weight 174.1
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluorosalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only