missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoropyridine, 98%

Catalog No. AAL1953906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1953906 5 g
AAL1953903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1953906 Supplier Thermo Scientific Chemicals Supplier No. L1953906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 71902-33-5 | C5H3F2N | 115.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71902-33-5
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.083
MDL Number MFCD03412222
InChI Key WRXAZPPGFLETFR-UHFFFAOYSA-N
Synonym pyridine, 3,5-difluoro, 3,5-difluoro-pyridine, abbypharma ap-14-5380, 3,5-difluoropyridine;, pubchem10556, acmc-209okl, 3,5-difluoro-pyridine;, ksc377a3n, 3,5-difluoropyridine, 3,5-difluoro pyridine
PubChem CID 642820
IUPAC Name 3,5-difluoropyridine
SMILES C1=C(C=NC=C1F)F

Specifications

Density 1.268
Boiling Point 92°C to 93°C
Flash Point 9°C (48°F)
Refractive Index 1.4437
Quantity 5 g
UN Number UN1993
Beilstein 7914363
Formula Weight 115.08
Percent Purity 98%
Chemical Name or Material 3,5-Difluoropyridine
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only