missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC427441000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC427441000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427441000 Supplier Thermo Scientific Chemicals Supplier No. 427441000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62351-47-7 | C6H3BrF2Mg | 217.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62351-47-7 , 109-99-9
Molecular Formula C6H3BrF2Mg
Molecular Weight (g/mol) 217.30
MDL Number MFCD00672005
InChI Key CURGXPMIRCWEDB-UHFFFAOYSA-M
Synonym 3,5-difluorophenylmagnesium bromide, bromo 3,5-difluorophenyl magnesium, 3,5-difluorophenylmagnesiumbromide, curgxpmircwedb-uhfffaoysa-m, 3,5-difluorophenyl magnesium bromide, 3,5-difluorophenylmagnesium bromide, 0.50m in 2-methf, 3,5-difluorophenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,5-difluorophenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
PubChem CID 2778376
IUPAC Name bromo(3,5-difluorophenyl)magnesium
SMILES FC1=CC([Mg]Br)=CC(F)=C1

Specifications

Color Yellow to brown solution
Boiling Point 65.0°C
CAS Min % 11.19, 86.57
CAS Max % 13.43, 88.81
Linear Formula F2C6H3MgBr
Concentration 0.5M, exact strength on the certificate of analysis
Density 0.9710g/mL
Formula Weight 217.29
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.971
Quantity 100 mL
Flash Point −17°C
Chemical Name or Material 3, 5-Difluorophenylmagnesium bromide, 0.5M solution in THF
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only