missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenol, 98+%

CAS: 2713-34-0 | C6H4F2O | 130.09 g/mol

$57.21 - $469.58

Chemical Identifiers

CAS 2713-34-0
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00002255
InChI Key HJSSBIMVTMYKPD-UHFFFAOYSA-N
PubChem CID 75928
IUPAC Name 3,5-difluorophenol
SMILES OC1=CC(F)=CC(F)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1517703
View Documents
Thermo Scientific Chemicals
A1517703
1 g
Each for $57.21
Only null left
 
AAA1517706
View Documents
Thermo Scientific Chemicals
A1517706
5 g
Each for $140.18
Only null left
 
AAA1517714
View Documents
Thermo Scientific Chemicals
A1517714
25 g
Each for $469.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2713-34-0
130.09
HJSSBIMVTMYKPD-UHFFFAOYSA-N
3,5-difluorophenol
C6H4F2O
MFCD00002255
75928
OC1=CC(F)=CC(F)=C1

Specifications

2713-34-0
65°C to 68°C (1 mmHg)
Phenol-like
MFCD00002255
UN3261
phenol, 3,5-difluoro, 3,5-difluoro phenol, phenol,3,5-difluoro, 3,5difluorophenol, 3,5-difluorphenol, 3.5-difluorophenol, 3,5-difluoro-phenol, phenol derivative, 6, pubchem1496, acmc-209gvc
OC1=CC(F)=CC(F)=C1
130.09
130.09
3,5-Difluorophenol
51°C to 55°C
70°C (158°F)
C6H4F2O
1 g
2078616
HJSSBIMVTMYKPD-UHFFFAOYSA-N
3,5-difluorophenol
75928
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H228-H302+H312+H332-H314-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only