missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenol, 98+%

CAS: 2713-34-0 | C6H4F2O | 130.09 g/mol

Supplier:  Thermo Scientific Chemicals A1517706

 View more versions of this product

Catalog No. AAA1517706


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Difluorophenol
51°C to 55°C
70°C (158°F)
C6H4F2O
5 g
2078616
HJSSBIMVTMYKPD-UHFFFAOYSA-N
3,5-difluorophenol
75928
≥98%
2713-34-0
65°C to 68°C (1 mmHg)
Phenol-like
MFCD00002255
UN3261
phenol, 3,5-difluoro, 3,5-difluoro phenol, phenol,3,5-difluoro, 3,5difluorophenol, 3,5-difluorphenol, 3.5-difluorophenol, 3,5-difluoro-phenol, phenol derivative, 6, pubchem1496, acmc-209gvc
OC1=CC(F)=CC(F)=C1
130.09
130.09

Chemical Identifiers

2713-34-0
130.09
HJSSBIMVTMYKPD-UHFFFAOYSA-N
3,5-difluorophenol
C6H4F2O
MFCD00002255
75928
OC1=CC(F)=CC(F)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H228-H302+H312+H332-H314-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only