missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoroanisole 97.0+%, TCI America™

Catalog No. D246310G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D246310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D246310G Supplier TCI America Supplier No. D246310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93343-10-3
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD00042560
InChI Key OTGQPYSISUUHAF-UHFFFAOYSA-N
Synonym 3,5-difluoroanisole, 3,5-difluoro-1-methoxybenzene, benzene, 1,3-difluoro-5-methoxy, 1,3-difluoro-5-methoxy-benzene, 3,5-difluoro anisole, 3,5-difluoroanisol, 3.5-difluoroanisole, pubchem1967, 3,5-difluoroanisole;, 3,5,-difluoroanisole;
PubChem CID 2724518
IUPAC Name 1,3-difluoro-5-methoxybenzene
SMILES COC1=CC(F)=CC(F)=C1

Specifications

Color Yellow
Boiling Point 148°C
Quantity 10 g
UN Number 1993
Formula Weight 144.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Difluoroanisole