missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenol 98.0+%, TCI America™

Catalog No. D036510G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D036510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D036510G Supplier TCI America Supplier No. D036510G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 591-35-5
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 162.997
MDL Number MFCD00002259
InChI Key VPOMSPZBQMDLTM-UHFFFAOYSA-N
Synonym phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
PubChem CID 11571
IUPAC Name 3,5-dichlorophenol
SMILES C1=C(C=C(C=C1Cl)Cl)O

Specifications

Melting Point 68°C
Boiling Point 122°C
Quantity 10 g
UN Number 2020
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorophenol
EINECSNumber (3)-0930
RTECSNumber SK8820000
TSCA Yes