missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzeneboronic acid, 98+%

Catalog No. AAB2276506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2276506 5 g
AAB2276503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2276506 Supplier Thermo Scientific Chemicals Supplier No. B2276506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67492-50-6 | C6H5BCl2O2 | 190.814 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67492-50-6
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.814
MDL Number MFCD00051935
InChI Key DKYRKAIKWFHQHM-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl boronic acid, 3,5-dichlorobenzeneboronic acid, 3,5-dichlorophenyl boranediol, 3,5-dichlophenylboronic acid, 3,5-dichloro phenylboronic acid, 3,5-dichlorobenzene boronic acid, 3,5-dichlorophenyl dihydroxyborane, boronic acid, b-3,5-dichlorophenyl, 3,5-dichlorophenylboronicacid
PubChem CID 2734331
IUPAC Name (3,5-dichlorophenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)Cl)Cl)(O)O

Specifications

Melting Point ∼315°C (decomposition)
Quantity 5 g
Beilstein 7581091
Formula Weight 190.82
Percent Purity ≥98%
Chemical Name or Material 3,5-Dichlorobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only