missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloroaniline 98,0+%, TCI America™

Catalog No. D0325500G
Change view
Click to view available options
Quantité:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
D0325500G 500 g
D032525G 25 g
D0325100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0325500G Supplier TCI America Supplier No. D0325500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-43-7
Formule moléculaire C6H5Cl2N
Poids moléculaire (g/mol) 162.013
Numéro MDL MFCD00007774
Clé InChI UQRLKWGPEVNVHT-UHFFFAOYSA-N
Synonyme benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline
PubChem CID 12281
ChEBI CHEBI:19904
Nom de l’IUPAC 3,5-dichloroaniline
SOURIRES C1=C(C=C(C=C1Cl)Cl)N

Specifications

Nom ou substance chimique 3,5-Dichloroaniline
Point de fusion 51 °C
Point d’ébullition 260 °C
Quantité 500 g
Numéro UN 3442
Poids de formule 162.01
Pourcentage de pureté ≥98,0% (GC)
Forme physique Poudre cristalline
EINECSNumber (3)-0261
TSCA Oui